CAS RN 5581-42-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Glyparamide, MedChemExpress
MedChemExpress Glyparamide is a chlorophenyl-containing sulfonylurea with hypoglycemic activity; Glyparamide rarely causes hepatic injury.