CAS RN 557-08-4
IUPAC Nombre:
zinc(2+) bis(undec-10-enoate)
Sinónimos:
zinc(2+) bis(undec-10-enoate)
Peso molecular (g/mol):
431.92
Fórmula molecular:
C22H38O4Zn
InChi Key:
YMCOHQVWOBMDCZ-UHFFFAOYSA-L
SMILES:
[Zn++].[O-]C(=O)CCCCCCCCC=C.[O-]C(=O)CCCCCCCCC=C
1
–
2
de
2
resultados
2
10-Undecenoic acid zinc salt, MedChemExpress
MedChemExpress 10-Undecenoic acid zinc salt is a natural or synthetic fungistatic fatty acid, is used topically in creams against fungal infections, eczemas, ringworm, and other cutaneous conditions.