CAS RN 55659-75-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress Deltonin
MedChemExpress Deltonin, a steroidal saponin, isolated from Dioscorea zingiberensis Wright, with antitumor activity; Deltonin inhibits ERK1/2 and AKT activation.