CAS RN 55395-07-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
IKarisoside A, MedChemExpress
MedChemExpress IKarisoside A (Icarisoside-A) is a natural flavonol glycoside and has anti-inflammatory properties.