CAS RN 552325-73-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
A-674563, MedChemExpress
MedChemExpress A-674563 is an orally active and selective Akt1 inhibitor with a Ki of 11 nM.