CAS RN 54197-31-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
R(+)-IAA-94, MedChemExpress
MedChemExpress R(+)-IAA-94 (R(+)-Methylindazone) is a potent indanyloxyacetic acid blocker of epithelial chloride channels. R(+)-IAA-94 inhibits Nef-sdAb19 (single-domain antibody) interaction and binds to negative factor (Nef).