CAS RN 537672-41-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
UF010, MedChemExpress
MedChemExpress UF010 is a potent and selective HDAC inhibitor with IC50 ∼0.06 μM, 0.1 μM, 0.5 μM and 1.5 μM for HDACs 3, 2, 1 and 8, respectively.