CAS RN 537034-17-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BML-210, MedChemExpress
MedChemExpress BML-210 is a novel HDAC inhibitor, and its mechanism of action has not been characterized.