CAS RN 524924-76-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Methoxy-PEPy, MedChemExpress
MedChemExpress Methoxy-PEPy is a potent and highly selective mGlu5 receptor antagonist with IC50 of 1 nM.