CAS RN 51550-25-5
IUPAC Nombre:
tetrasodium 4-hydroxy-5-{2-[(2E)-8-hydroxy-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}naphthalene-2,7-disulfonate
Sinónimos:
tetrasodium 4-hydroxy-5-{2-[(2E)-8-hydroxy-1-oxo-3,6-disulfonatonaphthalen-2-ylidene]hydrazin-1-yl}naphthalene-2,7-disulfonate
Peso molecular (g/mol):
738.5
Fórmula molecular:
C20H10N2Na4O15S4
InChi Key:
CBWXTVSARCWSMJ-QNKYWQQSSA-J
SMILES:
[Na+].[Na+].[Na+].[Na+].OC1=CC(=CC2=CC(=CC(N\N=C3/C(=O)C4=C(C=C(C=C4O)S([O-])(=O)=O)C=C3S([O-])(=O)=O)=C12)S([O-])(=O)=O)S([O-])(=O)=O
1
–
2
de
2
resultados
1
MedChemExpress Beryllon II
MedChemExpress Beryllon II is a widely used chromogenic reagent that is used to determine many elements, such as Mo, Mg and Co, and also used for the determination of proteins.