CAS RN 51415-02-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Chikusetsusaponin Iva, MedChemExpress
MedChemExpress Chikusetsusaponin IVa a major active ingredient of triterpenoid saponins, exerts antithrombotic effects, including minor hemorrhagic events.