CAS RN 503837-98-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Sortin1, MedChemExpress
MedChemExpress Sortin1 is a chemical genetic-hit molecule that causes specific mislocalization of plant and yeast-soluble and membrane vacuolar markers.