CAS RN 49843-98-3
IUPAC Nombre:
6-chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide
Sinónimos:
6-chloro-2,3,4,9-tetrahydro-1H-carbazole-1-carboxamide
Peso molecular (g/mol):
248.71
Fórmula molecular:
C13H13ClN2O
InChi Key:
FUZYTVDVLBBXDL-UHFFFAOYNA-N
SMILES:
NC(=O)C1CCCC2=C1NC1=CC=C(Cl)C=C21
1
–
4
de
4
resultados
2
Tocris Bioscience™ EX 527
CAS: 49843-98-3 Fórmula molecular: C13H13ClN2O
3
4
Selisistat, MedChemExpress
MedChemExpress Selisistat (EX-527) is a potent and selective SirT1 (Sir2 in Drosophila melanogaster) inhibitor with an IC50 of 123 nM for SirT1. Selisistat alleviates pathology in multiple animal and cell models of Huntington's disease.