CAS RN 486-62-4
IUPAC Nombre:
3-(4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4H-chromen-4-one
Sinónimos:
ononin formononetin glucoside
Peso molecular (g/mol):
430.41
Fórmula molecular:
C22H22O9
InChi Key:
MGJLSBDCWOSMHL-MIUGBVLSSA-N
SMILES:
COC1=CC=C(C=C1)C1=COC2=CC(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)=CC=C2C1=O
1
–
2
de
2
resultados
2
Ononin, MedChemExpress
MedChemExpress Ononin is an isoflavonoid, is an additional growth inhibitor in soils associated with the weed, Pluchea lanceolata.