CAS RN 483313-22-0
IUPAC Nombre:
2-({1-[5-(2-fluorophenyl)-2-methyl-1,3-thiazole-4-carbonyl]pyrrolidin-2-yl}methyl)-5-phenyl-1,3,4-oxadiazole
Sinónimos:
2-({1-[5-(2-fluorophenyl)-2-methyl-1,3-thiazole-4-carbonyl]pyrrolidin-2-yl}methyl)-5-phenyl-1,3,4-oxadiazole
Peso molecular (g/mol):
448.52
Fórmula molecular:
C24H21FN4O2S
InChi Key:
HYBZWVLPALMACV-UHFFFAOYNA-N
SMILES:
CC1=NC(C(=O)N2CCCC2CC2=NN=C(O2)C2=CC=CC=C2)=C(S1)C1=CC=CC=C1F
1
–
2
de
2
resultados
1
Tocris Bioscience™ SB 674042
CAS: 483313-22-0 Fórmula molecular: C24H21FN4O2S
2
SB-674042, MedChemExpress
MedChemExpress SB-674042 is a potent and selective non-peptide orexin OX1 receptor antagonist (Kd = 3.76 nM); exhibits 100-fold selectivity for OX1 over OX2 receptors.