CAS RN 480-43-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Isosakuranetin, MedChemExpress
MedChemExpress Isosakuranetin is a flavanone flavonoid which can be found in the fruit of Citrus bergamia.