CAS RN 477202-00-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Ipilimumab
MedChemExpress Ipilimumab is a fully human monoclonal IgG1κ antibody against the cytotoxic T-lymphocyte antigen-4 (CTLA-4), an immune-inhibitory molecule expressed in activated T cells and in suppressor T regulatory cells.
2
MedChemExpress Ipilimumab
MedChemExpress Ipilimumab is a fully human monoclonal IgG1κ antibody against the cytotoxic T-lymphocyte antigen-4 (CTLA-4), an immune-inhibitory molecule expressed in activated T cells and in suppressor T regulatory cells.