CAS RN 473728-58-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SCH 563705, MedChemExpress
MedChemExpress SCH 563705 is a potent and orally available CXCR2 and CXCR1 antagonist, with IC50s of 1.3 nM, 7.3 nM and Kis of 1 and 3 nM, respectively.