CAS RN 444723-13-1
IUPAC Nombre:
4-{[6-(cyclohexylmethoxy)-7H-purin-2-yl]amino}-N,N-diethylbenzamide
Sinónimos:
4-{[6-(cyclohexylmethoxy)-7H-purin-2-yl]amino}-N,N-diethylbenzamide
Peso molecular (g/mol):
422.53
Fórmula molecular:
C23H30N6O2
InChi Key:
XHEQSRJCJTWWAH-UHFFFAOYSA-N
SMILES:
CCN(CC)C(=O)C1=CC=C(NC2=NC(OCC3CCCCC3)=C3NC=NC3=N2)C=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ NU 6140
CAS: 444723-13-1 Fórmula molecular: C23H30N6O2
2
NU6140, MedChemExpress
MedChemExpress NU6140 is a selective CDK2-cyclin A inhibitor (IC50, 0.41 μM), exhibits 10- to 36-fold selectivity over other CDKs. NU6140 also potently inhibits Aurora A and Aurora B, with IC50s of 67 and 35 nM, respectively. Enhances the apoptotic effect, with anti-cancer activity.