CAS RN 440662-09-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ CeMMEC1
TAF1 inhibitor
2
CeMMEC1, MedChemExpress
MedChemExpress CeMMEC1 is an inhibitor of BRD4, and also has high affinity for TAF1, with an IC50 of 0.9 μM for TAF1, and a Kd of 1.8 μM for TAF1 (2).