CAS RN 439083-90-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Bay 60-7550, MedChemExpress
MedChemExpress Bay 60-7550 is a potent and selective PDE2 inhibitor with a Ki of 3.8 nM.