CAS RN 41945-48-6
IUPAC Nombre:
sodium 2-{2-[(4R)-4-[(1R,4R,5aS,7R,9aS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]acetamido}ethane-1-sulfonate
Sinónimos:
sodium 2-{2-[(4R)-4-[(1R,4R,5aS,7R,9aS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]acetamido}ethanesulfonate
Peso molecular (g/mol):
594.74
Fórmula molecular:
C28H47N2NaO8S
InChi Key:
PUODKHBECQMSKY-FQKBTHBQSA-M
SMILES:
[Na+].C[C@H](CCC(=O)NCC(=O)NCCS([O-])(=O)=O)[C@H]1CCC2C3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)C3C[C@H](O)[C@]12C
1
–
2
de
2
resultados
1
Sodium tauroglycocholate, MedChemExpress
MedChemExpress Sodium tauroglycocholate is an inhibitor of the biliary acid transporting system of the hepatocyte and also a surfactant used as a chemical permeation enhancer.