CAS RN 412950-08-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Rilapladib, MedChemExpress
MedChemExpress Rilapladib (SB 659032) is a selective Lp-PLA2 (lipoprotein-associated phospholipase A2) inhibitor with an IC50 of 230 pM. Rilapladib (SB 659032) is also a PAFR (Platelet Activating Factor Receptor) antagonist.