CAS RN 403811-55-2
IUPAC Nombre:
5-[(4-ethylphenyl)methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
Sinónimos:
5-[(4-ethylphenyl)methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
Peso molecular (g/mol):
249.35
Fórmula molecular:
C12H11NOS2
InChi Key:
SVXDHPADAXBMFB-UHFFFAOYSA-N
SMILES:
CCC1=CC=C(C=C2SC(=S)NC2=O)C=C1
1
–
3
de
3
resultados
2
R&D Systems™ 10058-F4
CAS: 403811-55-2 Fórmula molecular: C12H11NOS2
3
10058-F4, MedChemExpress
MedChemExpress 10058-F4 is a c-Myc inhibitor that prevents c-Myc-Max dimerization and transactivation of c-Myc target gene expression.