CAS RN 39011-91-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Oxypaeoniflorin, MedChemExpress
MedChemExpress Oxypaeoniflorin, an anti-oxidant, is a monoterpene glycoside compound isolated from Paeoniae species. Oxypaeoniflorin has neuroprotective and anti-inflammatory effects.