CAS RN 388592-44-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CK-869, MedChemExpress
MedChemExpress CK-869 is an Actin-Related Protein 2/3 (ARP2/3) complex inhibitor, with an IC50 of 7 μM.