CAS RN 38363-41-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(+)-Penbutolol, MedChemExpress
MedChemExpress (+)-Penbutolol is a β-adrenoceptor antagonist, with an IC50 of 0.74 μM. (+)-Penbutolol is an optical isomer of l-penbutolol with Na+ channel-blocking action.