CAS RN 383415-79-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Neuropeptide W-23(human)
MedChemExpress Neuropeptide W-23(human), the active form of Neuropeptide W, is an endogenous ligand for NPBW1 and NPBW2.
2
MedChemExpress Neuropeptide W-23(human)
MedChemExpress Neuropeptide W-23(human), the active form of Neuropeptide W, is an endogenous ligand for NPBW1 and NPBW2.