CAS RN 3820-67-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Glafenine, MedChemExpress
MedChemExpress Glafenine is a non-steroidal anti-inflammatory drug (NSAID), is a non-narcotic analgesic agent, widely used for the treatment of pains of various origins.