CAS RN 380607-77-2
IUPAC Nombre:
N-(2-{4-[2-(propane-2-sulfonamido)ethyl]phenyl}ethyl)propane-2-sulfonamide
Sinónimos:
N-(2-{4-[2-(propane-2-sulfonamido)ethyl]phenyl}ethyl)propane-2-sulfonamide
Peso molecular (g/mol):
376.53
Fórmula molecular:
C16H28N2O4S2
InChi Key:
FHLGMMYEKXPVSC-UHFFFAOYSA-N
SMILES:
CC(C)S(=O)(=O)NCCC1=CC=C(CCNS(=O)(=O)C(C)C)C=C1
1
–
1
de
1
resultados
1
CMPDA, MedChemExpress
MedChemExpress CMPDA is a positive allosteric modulator of AMPA receptors with EC50s of 45.4 (plusmn) 4.2 nM/63.4 (plusmn) 5.6 nM for GluA2i/GluA2o receptor.