CAS RN 37326-33-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Hyaluronidase, MedChemExpress
MedChemExpress Hyaluronidase (Hyaluronate 4-glycanohydrolase; Hyaluronoglucosaminidase) is a naturally occurring enzyme that depolymerizes hyaluronic acid by cleavage of glycosidic bonds and has been utilized in ophthalmic surgery.