CAS RN 37286-92-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Calcium polystyrene sulfonate, MedChemExpress
MedChemExpress Calcium polystyrene sulfonate is an ion-exchange resin used for reducing blood levels of potassium. Calcium polystyrene sulfonate is used to treat hyperkalemia in patients with chronic kidney disease (CKD).