CAS RN 372196-77-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PIK-75 hydrochloride, MedChemExpress
MedChemExpress PIK-75 hydrochloride is a reversible DNA-PK and p110α-selective inhibitor, which inhibits DNA-PK, p110α and p110γ with IC50s of 2, 5.8 and 76 nM, respectively. PIK-75 hydrochloride inhibits p110α >200-fold more potently than p110β (IC50=1.3 μM). PIK-75 hydrochloride induces apoptosis.