CAS RN 371942-69-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
YM-201636, MedChemExpress
MedChemExpress YM-201636 is a potent and selective PIKfyve inhibitor with an IC50 of 33 nM. YM-201636 also inhibits p110α with an IC50 of 3.3 μM. YM-201636 inhibits retroviral replication.