CAS RN 348086-71-5
IUPAC Nombre:
N-methyl-N-(4-methyl-5-sulfamoyl-1,3-thiazol-2-yl)-2-[4-(pyridin-2-yl)phenyl]acetamide
Sinónimos:
N-methyl-N-(4-methyl-5-sulfamoyl-1,3-thiazol-2-yl)-2-[4-(pyridin-2-yl)phenyl]acetamide
Peso molecular (g/mol):
402.49
Fórmula molecular:
C18H18N4O3S2
InChi Key:
IVZKZONQVYTCKC-UHFFFAOYSA-N
SMILES:
CN(C(=O)CC1=CC=C(C=C1)C1=CC=CC=N1)C1=NC(C)=C(S1)S(N)(=O)=O
1
–
3
de
3
resultados
2
Tocris Bioscience™ Pritelivir
Potent and selective herpes simplex virus (HSV) helicase primase inhibitor
3
Pritelivir, MedChemExpress
MedChemExpress Pritelivir (AIC316), an inhibitor of the viral helicase-primase complex, exhibits antiviral activity in vitro and in animal models of herpes simplex virus (HSV) infection. Pritelivir is active against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2) with the IC50 of 0.02 μM against HSV1-2.