CAS RN 34420-19-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Lathyrol
MedChemExpress Lathyrol is a natural product, and is used for cancer treatment.
2
MedChemExpress Lathyrol
MedChemExpress Lathyrol is a natural product, and is used for cancer treatment.
3
MedChemExpress Lathyrol
MedChemExpress Lathyrol is a natural product, and is used for cancer treatment.