CAS RN 34215-57-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress 3,3'-Dioctadecyloxacarbocyanine perchlorate
MedChemExpress 3,3′-Dioctadecyloxacarbocyanine perchlorate is a green fluorescent lipophilic tracer, which is weakly fluorescent in water but highly fluorescent and quite photostable when incorporated into membranes.