CAS RN 3397-23-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
4
de
4
resultados
1
MedChemExpress Ornipressin
MedChemExpress Ornipressin is a potent vasoconstrictor, hemostatic and renal agent.
2
MedChemExpress Ornipressin
MedChemExpress Ornipressin is a potent vasoconstrictor, hemostatic and renal agent.
3
MedChemExpress Ornipressin
MedChemExpress Ornipressin is a potent vasoconstrictor, hemostatic and renal agent.
4
MedChemExpress Ornipressin
MedChemExpress Ornipressin is a potent vasoconstrictor, hemostatic and renal agent.