CAS RN 335-58-0
IUPAC Nombre:
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-pentadecafluoro-7-iodoheptane
Sinónimos:
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-pentadecafluoro-7-iodoheptane
Peso molecular (g/mol):
495.96
Fórmula molecular:
C7F15I
InChi Key:
AHUMDLIBMIYQMU-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
1
–
1
de
1
resultados
1
Perfluoroheptil Yoduro, TRC
CAS: 335-58-0 Fórmula molecular: C7F15I Peso molecular (g/mol): 495.96 Sinónimo: 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-Pentadecafluoro-7-iodoheptane,Pentadecafluoro-1-iodoheptane,1-Iodoperfluoroheptane,Pentadecafluoro-1-iodoheptane,Pentadecafluoroheptyl Iodide,Perfluoro-1-iodoheptane,Perfluoro-n-heptyl Iodide Nombre IUPAC: 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-pentadecafluoro-7-iodoheptano SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I