CAS RN 334969-03-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Recilisib, MedChemExpress
MedChemExpress Recilisib (ON 01210) is a radioprotectant, which can activate AKT, PI3K activities in cells.