CAS RN 334951-92-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MEK inhibitor, MedChemExpress
MedChemExpress MEK inhibitor is a potent MEK inhibitor with antitumor potency.