CAS RN 333-36-8
IUPAC Nombre:
1,1,1-trifluoro-2-(2,2,2-trifluoroethoxy)ethane
Sinónimos:
flurothyl hexafluorodiethyl ether flurothyl (substance) fluorothyl flurotyl 氟替尔 flurotylum flurotilo indoklon
Peso molecular (g/mol):
182.07
Fórmula molecular:
C4H4F6O
InChi Key:
KGPPDNUWZNWPSI-UHFFFAOYSA-N
SMILES:
FC(F)(F)COCC(F)(F)F
1
–
1
de
1
resultados
1