CAS RN 33289-85-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Dipsacoside B, MedChemExpress
MedChemExpress Dipsacoside B is a major bioactive saponin, which can be used as a marker.