CAS RN 33231-14-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DZ2002, MedChemExpress
MedChemExpress DZ2002 is a potent and reversible S-Adenosyl-L-homocysteine Hydrolase(SAHH; AdoHcy Hydrolase) inhibitor with Ki of 17.9 nM.