CAS RN 332164-34-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HFI-142, MedChemExpress
MedChemExpress HFI-142 is an insulin-regulated aminopeptidase (IRAP) inhibitor with a Ki of 2.01 μM.