CAS RN 330676-02-3
IUPAC Nombre:
2-(1H-1,3-benzodiazol-2-ylsulfanyl)-N'-[(3-bromo-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]propanehydrazide
Sinónimos:
2-(1H-1,3-benzodiazol-2-ylsulfanyl)-N'-[(3-bromo-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]propanehydrazide
Peso molecular (g/mol):
419.3
Fórmula molecular:
C17H15BrN4O2S
InChi Key:
GEHVZUUHMWNMAY-UHFFFAOYNA-N
SMILES:
CC(SC1=NC2=CC=CC=C2N1)C(=O)NNC=C1C=C(Br)C=CC1=O
1
–
2
de
2
resultados
1
Tocris Bioscience™ KH 7
CAS: 330676-02-3 Fórmula molecular: C17H15BrN4O2S
2
KH7, MedChemExpress
MedChemExpress KH7 is a soluble adenylyl cyclase (sAC)-specific inhibitor, with IC50s of 3-10 μM toward both recombinant purified human sACt protein and heterologously expressed sACt in cellular assays. KH7 is also a cAMP inhibitor.