CAS RN 330161-87-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ SU 6656
Potent ATP-competitive Src family kinase inhibitor (IC50 values are 0.02, 0.13, 0.17, 0.28 and 6.88 nM for Yes, Lyn, Fyn, Src and Lck, respectively)
2
SU6656, MedChemExpress
MedChemExpress SU6656 is a Src family kinases inhibitor with IC50s of 280, 20, 130, 170 nM for Src, Yes, Lyn, and Fyn, respectively. SU6656 inhibits FAK phosphorylation at Y576/577, Y925, Y861 sites. SU6656 also inhibits p-AKT.