CAS RN 329059-55-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BRD7116, MedChemExpress
MedChemExpress BRD7116 competitively binds to bacterial DNA gyrase, exhibits an EC50 of 200 nM for LSCe cells, with cell-non-autonomous anti-leukemia activity.