CAS RN 328541-79-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ GlyH 101
Reversible, voltage-dependent CFTR chloride channel blocker
2
GlyH-101, MedChemExpress
MedChemExpress GlyH-101 is a cell-permeable glycinyl hydrazone compound that blocks CFTR with Ki of 1.4 uM.
3