Learn More
Tocris Bioscience™ GlyH 101
Descripción
GlyH 101 is a reversible, voltage-dependent CFTR chloride channel blocker (Ki = 4.3 μM). Inhibits forskolin -induced hyperpolarization in nasal potential differences and inhibits cholera toxin-induced intestinal fluid secretion in mice.
Especificaciones
Especificaciones
| Nombre del producto químico o material | GlyH 101 |
| CAS | 328541-79-3 |
| Sinónimo | N-2-Naphthalenyl-2-[(3,5-dibromo-2,4-dihydroxyphenyl)methylene]glycine hydrazide |
| Cantidad | 50 mg |
| Solubilidad | Soluble to 100mM in DMSO |
| Fórmula molecular | C19H15Br2N3O3 |
| Formula Weight (peso de la fórmula) | 493.15 |
| Forma física | Sólido |
| Tipo de producto | Bioactive Small Molecule |
| Pureza | >98% |
Research Use Only. Not For Human or Veterinary Use